Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB01031

Sigma-Aldrich

Antheindurolide A

≥90% (LC/MS-ELSD)

Synonym(s):

2(3H)-Furanone, dihydro-3-methylene-4-[3-methyl-4-(tetrahydro-4-methyl-5-oxo-2-furanyl)-2-butenyl]- (9CI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O4
CAS Number:
Molecular Weight:
264.32
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

264.32

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1C[C@@H](C(=C)C1=O)C\C=C(\CC2OC(=O)C(C2)C)/C

InChI key

KFKSQOIZZRZEFC-BZFAMWISSA-N

General description

Antheindurolide A, a sesquiterpene lactone, is commonly found in Anthemis indurata, Anthemis arvensis, and Anthemis cotula.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, cell signaling, metabolomics and phytochemical research.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service