Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00573

Sigma-Aldrich

Zataroside B

≥90% (LC/MS-UV)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24O7
CAS Number:
Molecular Weight:
328.36
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥90% (LC/MS-UV)

form

solid

solubility

DMSO: 1 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O[C@@H]([C@@H](O[C@@H]([C@H]1O)CO)OC2=C(C)C=C(O)C(C(C)C)=C2)[C@H]1O

InChI

1S/C16H24O7/c1-7(2)9-5-11(8(3)4-10(9)18)22-16-15(21)14(20)13(19)12(6-17)23-16/h4-5,7,12-21H,6H2,1-3H3/t12-,13-,14+,15-,16-/m1/s1

InChI key

XFPSRAYOBUHKRV-IBEHDNSVSA-N

General description

Natural product derived from plant source.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service