Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00413

Sigma-Aldrich

α-GalNAc-PEG3-Alkyne

≥95% (HPLC)

Synonym(s):

α-GalNAc-PEG3-Propyne, (9-Propargyl-3,6,9-trioxadodecyl) 2-acetamido-2-deoxy-α-D-galactopyranoside, (Dodec-11-yn-3,6,9-trioxadodecyl) 2-acetamido-2-deoxy-α-D-galactopyranoside, 2-{2-[2-(Prop-2-ynyloxy)ethoxy]ethoxy}ethyl 2-acetamido-2-deoxy-α-D-galactopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H29NO9
Molecular Weight:
391.41
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥95% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H](CO)O[C@H](OCCOCCOCCOCC#C)[C@@H]([C@H]1O)NC(C)=O

InChI

1S/C17H29NO9/c1-3-4-23-5-6-24-7-8-25-9-10-26-17-14(18-12(2)20)16(22)15(21)13(11-19)27-17/h1,13-17,19,21-22H,4-11H2,2H3,(H,18,20)/t13?,14?,15-,16+,17-/m0/s1

InChI key

NKSPHILIVROOQP-ZKLCHKPXSA-N

Biochem/physiol Actions

α-GalNAc-PEG3-Alkyne is a PEGylated glycoside. PEGylated glycosides are used as ligands for Click Chemistry. They may also be important for use as carbohydrate carriers in glycotherapeutic applications, drug carriers for site specific delivery, drug enhancers for decreasing toxicity and solubility and protein enhancers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service