Skip to Content
MilliporeSigma
All Photos(1)

Documents

SMB00353

Sigma-Aldrich

N-Methylcytisine

≥98% (HPLC)

Synonym(s):

(1R)-1,2,3,4,5,6-hexahydro-3-methyl-1,5-methano-8H-pyrido(1,2a)(1,5)diazocin-8-one, Caulophylline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16N2O
CAS Number:
Molecular Weight:
204.27
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

CN1C[C@@H]2C[C@H](C1)C3=CC=CC(=O)N3C2

InChI

1S/C12H16N2O/c1-13-6-9-5-10(8-13)11-3-2-4-12(15)14(11)7-9/h2-4,9-10H,5-8H2,1H3/t9-,10+/m0/s1

InChI key

CULUKMPMGVXCEI-VHSXEESVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Methylcytisine is a nicotinic alkaloid found in Caulophyllum thalictroides, also known as blue cohosh.

Biochem/physiol Actions

N-Methylcytisine is toxic and binds selectively to nicotinic acetylcholine receptors in the CNS.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service