Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00347

Sigma-Aldrich

Ginsenoside F1

≥98% (HPLC)

Synonym(s):

Ginsenoside F1, (3β,6α,12β)-3,6,12-trihydroxydammar-24-en-20-yl β-D-glucopyranoside, 20(S)-ginsenoside F1, panaxoside A progenin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H62O9
CAS Number:
Molecular Weight:
638.87
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

O[C@H]1CC[C@@]2(C)[C@]([C@@H](O)C[C@]3(C)[C@]2([H])C[C@@H](O)[C@@]4([H])[C@@]3(C)CC[C@]4([H])[C@](C)(CCC=C(C)C)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)([H])C1(C)C

InChI

1S/C36H62O9/c1-19(2)10-9-13-36(8,45-31-29(43)28(42)27(41)23(18-37)44-31)20-11-15-34(6)26(20)21(38)16-24-33(5)14-12-25(40)32(3,4)30(33)22(39)17-35(24,34)7/h10,20-31,37-43H,9,11-18H2,1-8H3/t20-,21+,22-,23+,24+,25-,26-,27+,28-,29+,30-,31-,33+,34+,35+,36-/m0/s1

InChI key

XNGXWSFSJIQMNC-FIYORUNESA-N

General description

Ginsenoside F1 is a saponin found in the roots of Panax ginseng.

Biochem/physiol Actions

Ginsenoside F1 is an active ingredient of ginseng and shows anti-aging and antioxidation properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service