Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCP0030

Sigma-Aldrich

[Gly22]-Amyloid β 1-42 Arctic human

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C200H307N55O58S1
Molecular Weight:
4441.98
UNSPSC Code:
12352209
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... APP(351)

Amino Acid Sequence

Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Gly-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala

General description

β-Amyloid 1-42 is one of the major constituents of the neurofibrillary tangles and plaques characteristic of the Alzheimer brain. The Arctic mutation E22G causes early onset of Alzheimer′s compared to wild type and promotes protofibril formation and neurotoxicity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiujian Yu et al.
Biological research, 51(1), 35-35 (2018-09-21)
The previous studies have demonstrated the reduction of thiamine diphosphate is specific to Alzheimer's disease (AD) and causal factor of brain glucose hypometabolism, which is considered as a neurodegenerative index of AD and closely correlates with the degree of cognitive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service