Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SBR00045

Sigma-Aldrich

Gentamicin-Amphotericin B Ready Made Solution

Gentamicin 25mg/mL and Amphotericin B 125µg/mL

Synonym(s):

Gentamicin-Amphotericin B Ready Made Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51281607
NACRES:
NA.76

Quality Level

specific activity

Antibacterial and antifungal

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
fungi
mycoplasma
yeast

Mode of action

cell membrane | interferes
protein synthesis | interferes

storage temp.

−20°C

General description

Gentamicin-Amphotericin B is an aqueous based stable formulation, intended to eradicate sensitive bacterial and fungal infections. Gentamicin is an aminoglycoside class antibiotic inhibiting the protein synthesis preventing the growth of gram positive and gram-negative bacteria. Amphotericin B is a well-known polyene antifungal compound used in cell culture. The Gentamicin-Amphotericin B mixture is especially usable for cell culture applications and as part of viral transfer media (VTM).

Other Notes

  • The Gentamicin-Amphotericin B mixture is composed from Gentamicin 25mg/ml and Amphotericin B 125 µg/mL.
  • The working concentrations according to CDC SOP# DSR-052-05 are 100 µg/mL for Gentamicin and 0.5 µg/mL for Amphotericin B.
  • 2mL Gentamicin-Amphotericin B stock mixture is sufficient to prepare 500 mL working concentration.
  • The product is packed under aseptic conditions.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Criticality of in-house preparation of viral transport medium in times of shortage during COVID-19 pandemic
Petersen, Jeffrey, Sharvari Dalal, et al
Laboratory Medicine, 52, no. 2, e39-e45 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service