Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.56
UNSPSC Code:
12352203
Conjugate:
magnetic beads
Species reactivity:
-
Application:
—
Citations:
3
General description
Anti-HA Magnetic Beads are a conjugate of high-affinity mouse IgG1 monoclonal anti-HA antibody against HA tag (YPYDVPDYA) derived from human influenza the hemagglutinin (HA) protein. The high degree of charged amino acids in the HA-tag enhances its potential for strong antibody recognition. This product may be used for purification of HA-tagged proteins that are expressed in bacterial and mammalian cell lysates.
Application
These anti-HA magnetic agarose beads may be used in various immunological techniques, such as immunoprecipitation, immunoaffinity purification, ubiquitination assay.
Features and Benefits
- Antibody is conjugated to cyanogen bromide-activated magnetic agarose beads.
- Protein-to-beads ratio ~ 4 mg/mL.
- High specificity binding to HA-tagged proteins.
Other Notes
This product is for R&D use only. Not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Storage Class
12 - Non Combustible Liquids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.