Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5700598

Sigma-Aldrich

Recombinant 2019-nCoV 3C-like Proteinase with His and Avi tag

Synonym(s):

3C-like main protease, 3CL Proteinase, 3CL-Mpro, M Proteinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.26

recombinant

expressed in E. coli

Quality Level

Assay

>95% (SDS-PAGE)

form

liquid

mol wt

36.9 kDa

impurities

<1.0 EU/μg protein (LAL test)

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

SARS virus ... ORF1ab(43740578)

General description

Recombinant 2019-nCoV 3C-like Proteinase is produced by E. coli expression system. The target protein is expressed with sequence (Ser1-Gln306) of 2019-ncov 3C-like Proteinase (Accession #YP_009725301.1) fused with an initial Met, a 6XHis, Avi tag at the N-terminus.

Storage and Stability

This product is stable at ≤ -70°C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store at recommended temperature. Avoid repeated freeze-thaw cycles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Data presented is the available current product information and provided as-is. These products have not been tested or verified in any additional applications, sample types, including any clinical use. Experimental conditions must be empirically derived by the user. Our Antibody Guarantee only covers tested applications stated herein and conditions presented in our product information and is not extended to publications.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service