Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB5600268

Sigma-Aldrich

Anti-Terminal deoxynucleotidyl transferase (TdT) antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM379, purified immunoglobulin

Synonym(s):

DNA nucleotidylexotransferase, DNTT, Tdt

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

recombinant

expressed in HEK 293 cells

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM379, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:100—1:200
western blot: 1:1000-1:2000

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DNTT(1791)

Specificity

This antibody reacts to human Terminal deoxynucleotidyl transferase (TdT).

Immunogen

A peptide corresponding to human Terminal deoxynucleotidyl transferase (TdT)

Physical form

Solution in phosphate buffered saline containing 50% Glycerol/PBS with 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamara Aulia Fakhrinnisa et al.
Asian Pacific journal of cancer prevention : APJCP, 22(2), 457-462 (2021-03-01)
Cisplatin is potent chemotherapy for broad-spectrum malignancies treatment, but its use is limited by organ toxicity effects, including nephrotoxicity. Glutamine prevents cisplatin nephrotoxicity by inhibiting the oxidative stress in kidney cell apoptosis. This research examined the nephroprotective effects of intravenous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service