Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB5600195

Sigma-Aldrich

Anti-Mouse IgG antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM104, purified immunoglobulin

Synonym(s):

Mouse Immunoglobulin G

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

recombinant

expressed in HEK 293 cells

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM104, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

mouse

technique(s)

ELISA: 0.005-0.2 μg/mL
immunocytochemistry: 0.5-2 μg/mL
immunohistochemistry: 0.5-2 μg/mL
immunoprecipitation (IP): suitable (assay dependent concentration)
western blot: 0.1— 0.5 μg/mL

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Specificity

This antibody reacts to the Fc region of all subclasses of mouse IgG. No cross reactivity with mouse IgM, IgA, IgE, human IgG, and rat IgG. It may cross react to goat IgG.

Immunogen

Mouse IgG

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 50% Glycerol/PBS with 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jincheng Liu et al.
Stem cells translational medicine, 12(5), 307-321 (2023-04-04)
N6-methyladenosine (m6A) methylation, a well-known modification with new epigenetic functions, has been reported to participate in the progression of osteoporosis (OP), providing novel insights into the pathogenesis of OP. However, as the key component of m6A methylation, Wilms tumor 1-associated
Zhiyong Ren et al.
Oncology letters, 24(5), 388-388 (2022-10-25)
The aim of this study was to investigate the potential mechanisms of long noncoding (lnc) RNA Just proximal to X-inactive specific transcript (JPX) in interleukin (IL)-1β-stimulated chondrocytes. Human C28/I2 chondrocytes were treated with IL-1β to simulate osteoarthritic (OA) injury. The
Michael J Scherm et al.
Cell reports, 41(4), 111562-111562 (2022-10-27)
Infection by the Ebola virus, a member of the Filoviridae family of RNA viruses, leads to acute viral hemorrhagic fever. End-stage Ebola virus disease is characterized by a cytokine storm that causes tissue damage, vascular disintegration, and multi-organ failure. Previous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service