Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB5600134

Sigma-Aldrich

Anti-Acetyl-alpha-Tubulin (Lys40) antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM318, purified immunoglobulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

recombinant

expressed in HEK 293 cells

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM318, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

vertebrates

technique(s)

immunofluorescence: 1:1000-1:5000

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

acetylation (Lys40)

Gene Information

human ... TUBA1A(7846)

Specificity

This antibody reacts to alpha-Tubulin acetylated at Lysine 40. No cross reactivity to non-acetylated alpha-Tubulin at Lysine 40

Immunogen

An acetyl-peptide corresponding to Acetyl- alpha-Tubulin (Lys40)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 50% glycerol, 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quanchao Sun et al.
Experimental lung research, 46(10), 393-408 (2020-10-13)
3 D imaging of the lung is not a trivial undertaking as during preparation the lung may collapse. Also serial sections and scans followed by 3 D reconstruction may lead to artifacts. The present study aims to figure out the best way
Pamela Meneses Iack et al.
Journal of neurochemistry, 161(4), 320-334 (2021-12-24)
Neocortex development comprises of a complex series of time- and space-specific processes to generate the typical interconnected six-layered architecture of adult mammals. Axon growth is required for the proper establishment of cortical circuits. Malformations in axonal growth and pathfinding might
Anna Verschueren et al.
Communications biology, 5(1), 89-89 (2022-01-26)
Human cone phototropism is a key mechanism underlying the Stiles-Crawford effect, a psychophysiological phenomenon according to which photoreceptor outer/inner segments are aligned along with the direction of incoming light. However, such photomechanical movements of photoreceptors remain elusive in mammals. We
Jingjing Pei et al.
Cell reports, 37(2), 109803-109803 (2021-10-14)
Human respiratory syncytial virus (RSV) is a common cause of lower respiratory tract infections in the pediatric, elderly, and immunocompromised individuals. RSV non-structural protein NS1 is a known cytosolic immune antagonist, but how NS1 modulates host responses remains poorly defined.
Zhiyuan You et al.
Nature communications, 10(1), 5792-5792 (2019-12-21)
Autophagy receptor p62/SQSTM1 promotes the assembly and removal of ubiquitylated proteins by forming p62 bodies and mediating their encapsulation in autophagosomes. Here we show that under nutrient-deficient conditions, cellular p62 specifically undergoes acetylation, which is required for the formation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service