Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB5600005

Sigma-Aldrich

Anti-Human IgG2 antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM118, purified immunoglobulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

recombinant

expressed in HEK 293 cells

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM118, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: suitable
immunocytochemistry: 0.5-2 μg/mL
immunohistochemistry: 0.5-2 μg/mL

isotype

IgG

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Specificity

This antibody reacts to the heavy chain of human IgG2. No cross reactivity with human IgG1, IgG3, IgG4, IgM, IgA, IgD, IgE, mouse IgG, rat IgG, or goat IgG.

Immunogen

Purified human IgG2 whole molecule

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 50% glycerol, 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service