Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB5500142

Sigma-Aldrich

Anti-MCM2 antibody, Rabbit monoclonal

clone SP85, recombinant, expressed in proprietary host, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SP85, monoclonal

species reactivity

human (tested)

species reactivity (predicted by homology)

frog, bovine, mouse, rat, dog

technique(s)

flow cytometry: 1:100
immunoblotting: 1:100
immunohistochemistry: 1:100

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... MCM2(4171)

General description

Mini chromosome maintenance protein 2 (MCM2) is a human nuclear protein. This cell-cycle regulatory protein is similar to members of the family of early S-phase proteins and is expressed in four phases of the cell cycle. MCM2 gene is mapped to human chromosome 3q21.3. MCM2 protein is a member of eukaryal MCM2–7 subunits and is expressed in the nucleus and cytoplasm of various tissues.

Immunogen

Synthetic peptide derived from internal region of human MCM2 protein.

Biochem/physiol Actions

Mini chromosome maintenance protein 2 (MCM2) is involved in regulating DNA replication, is crucial in the cell cycle. From its localization, CDCL1 is a candidate for an oncogene affected by chromosomal breaks in acute myeloid leukemia (AML). MCM-2 levels are elevated in salivary gland tumors and malignant lesions of the oral cavity. It serves as a marker for the detection of oral squamous cell carcinoma (OSCC). MCM2 overexpression also displays pro-apoptotic and apoptotic. MCM2 is also implicated in nonsyndromic hearing loss.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

François-Xavier Nouhaud et al.
Clinical genitourinary cancer, 16(4), e795-e805 (2018-03-20)
Gene copy number variations (CNVs) have been reported to be frequent in renal cell carcinoma (RCC), with potential prognostic value for some. However, their clinical utility, especially to guide treatment of metastatic disease remains to be established. Our objectives were
Petra Mlcochova et al.
The EMBO journal, 37(1), 50-62 (2017-11-01)
We report that DNA damage induced by topoisomerase inhibitors, including etoposide (ETO), results in a potent block to HIV-1 infection in human monocyte-derived macrophages (MDM). SAMHD1 suppresses viral reverse transcription (RT) through depletion of cellular dNTPs but is naturally switched
A Mincheva et al.
Cytogenetics and cell genetics, 65(4), 276-277 (1994-01-01)
BM28, a newly recognized human nuclear protein, possibly plays an important role in two crucial steps of the cell cycle e.g. the onset of DNA replication and cell division. It shows significant similarity to members of a recently defined family
Seyed Mohammad Razavi et al.
The Malaysian journal of pathology, 37(3), 253-258 (2015-12-30)
Proteins necessary for DNA replication and normal regulation for the cell cycle include minichromosome maintenance-2 (Mcm-2). Overexpression of this protein in several premalignant and malignant lesions has been observed. In this study, the diagnostic value of Mcm-2 expression in distinguishing
Petra Mlcochova et al.
The EMBO journal, 36(5), 604-616 (2017-01-27)
An unresolved question is how HIV-1 achieves efficient replication in terminally differentiated macrophages despite the restriction factor SAMHD1. We reveal inducible changes in expression of cell cycle-associated proteins including MCM2 and cyclins A, E, D1/D3 in macrophages, without evidence for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service