Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5500118

Sigma-Aldrich

Anti-HLA-DP alpha 1 antibody, Rabbit monoclonal

recombinant, expressed in proprietary host, clone SP228, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SP228, monoclonal

species reactivity

human (tested)

technique(s)

flow cytometry: 1:100
immunohistochemistry: 1:100

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... HLA-DPA1(3113)

General description

HLA-DP is an antigen receptor and one of the MHC class II proteins. HLA-DP is a heterodimer of an alpha and a beta chain. Both chains are type I transmembrane proteins with short cytoplasmic domains. It is expressed on antigen-presenting cells and a variety of other cell types, and is also expressed by various tumors such as melanoma, gastric carcinoma, breast carcinoma, ovarian carcinoma, and lymphoma.
Major histocompatibility complex, class II, DP alpha 1 (HLA-DPA1) is a class II heterodimer comprising of a polymorphic β and an oligomorphic α chain. It is encoded by the gene mapped to human chromosome 6p21.33. HLA-DPA1 is generally expressed on a subset of Kupffer cells and resident macrophages in liver.

Immunogen

Synthetic peptide derived from human HLA-DP alpha1 protein.

Application

Anti-HLA-DP alpha 1 antibody, Rabbit monoclonal has been used in immunohistochemistry.

Biochem/physiol Actions

Major histocompatibility complex, class II, DP alpha 1 (HLA-DPA1) alloreactivity is highly relevant in clinical transplantation. It plays an essential role in the presentation of viral antigens to CD4+ T helper (Th) lymphocytes. Studies have suggested that SNP (single nucleotide polymorphisms) of HLA-DP can result in chronic hepatitis B virus (HBV) infection.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pulsed electrical stimulation benefits wound healing by activating skin fibroblasts through the TGF?1/ERK/NF-?B axis.
Wang Y, et al.
Biochimica et Biophysica Acta, 1860(7), 1551-1559 (2016)
HLA-DP Polymorphisms Affect the Outcomes of Chronic Hepatitis B Virus Infections, Possibly through Interacting with Viral Mutations
Zhang Q, et al.
Journal of Virology, 02073-02073 (2013)
Allorecognition of HLA-DP by CD4+ T cells is affected by polymorphism in its alpha chain.
Lauterbach N, et al.
Molecular Immunology, 59(1), 19-29 (2014)
Studies on the association of single nucleotide polymorphisms of HLA-DP and DQ genes with the outcome of chronic hepatitis B virus infection
Su M, et al.
Zhonghua yi xue yi chuan xue za zhi (Chinese Journal of Medical Genetics), 31(6), 765-769 (2014)
New loci associated with chronic hepatitis B virus infection in Han Chinese.
Hu Z, et al.
Nature Genetics, 45(12), 1499-1499 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service