Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB5500037

Sigma-Aldrich

Anti-CD13 antibody, Rabbit monoclonal

clone SP182, recombinant, expressed in proprietary host, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SP182, monoclonal

species reactivity

human (tested)

species reactivity (predicted by homology)

rabbit, pig

technique(s)

flow cytometry: 1:400
immunohistochemistry: 1:100

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ANPEP(290)

Related Categories

General description

CD13, also known as aminopeptidase N, participates in the final digestion of peptides from stomach, pancreas, and other organs. It expresses by diverse cell types including small intestinal and tubular epithelial cells, macrophages, granulocytes and synaptic membranes. It may be involved in the cleavage of antigen peptides, degradation of neurotransmitters, and angiogenesis. It is one of the useful markers for acute myeloid leukemia or lymphoma.

Immunogen

Synthetic peptide derived from the internal region of human CD13 protein.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service