Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5500014

Sigma-Aldrich

Anti-BCL-2 Alpha antibody, Rabbit monoclonal

recombinant, expressed in proprietary host, clone SP66, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
FACS
IHC
clone:
SP66, monoclonal
species reactivity:
human (tested)
citations:
4
technique(s):
flow cytometry: 1:100
immunohistochemistry: 1:100

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SP66, monoclonal

species reactivity

human (tested)

technique(s)

flow cytometry: 1:100
immunohistochemistry: 1:100

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... BCL2(596)

Related Categories

General description

B-cell lymphoma 2 (BCL-2) is an anti-apoptotic and target gene of nuclear factor-κB (NF-κB). The protein is expressed in the nuclear envelope, mitochondria and endoplasmic reticulum. The gene encoding BCL-2 is localized on human chromosome 18q21.

Immunogen

Synthetic peptide corresponding to N-terminus of human BCL-2 alpha protein.

Biochem/physiol Actions

B-cell lymphoma 2 (BCL-2) inhibits apoptosis in cells and has been shown to be upregulated in adenomas. Expression of BCL-2α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of BCL-2α protein, whereas the normal or hyperplastic germinal centers are negative.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bcl-2 localized at the nuclear compartment induces apoptosis after transient overexpression.
Portier BP and Taglialatela G
The Journal of Biological Chemistry, 281(52), 40493-40502 (2006)
Structure-based lead optimization and biological evaluation of BAX direct activators as novel potential anticancer agents.
Stornaiuolo M
Journal of Medicinal Chemistry, 58(5), 2135-2148 (2015)
Xiaopeng Wang et al.
Molecular medicine reports, 23(2) (2020-12-11)
Acute progressive hypoxic respiratory failure caused by various predisposing factors is known as acute respiratory distress syndrome (ARDS). Although penehyclidine hydrochloride (PHC), an anticholinergic drug, is widely applied in clinical practice, the specific mechanisms underlying PHC in the treatment of
Maartje van der Heijden et al.
Nature communications, 7, 10916-10916 (2016-03-10)
Intestinal tumour formation is generally thought to occur following mutational events in the stem cell pool. However, active NF-κB signalling additionally facilitates malignant transformation of differentiated cells. We hypothesized that genes shared between NF-κB and intestinal stem cell (ISCs) signatures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service