Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5300249

Sigma-Aldrich

Monoclonal Anti-MLH1 antibody produced in mouse

clone 4C9C7, ascites fluid

Synonym(s):

COCA2, FCC2, HNPCC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

4C9C7, monoclonal

mol wt

85 kDa

species reactivity

human, monkey

technique(s)

direct ELISA: 1:10,000
immunohistochemistry: 1:200-1:1,000
indirect immunofluorescence: 1:200-1:1,000
western blot: 1:500-1:2,000

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MLH1(4292)

Immunogen

Purified recombinant fragment of MLH1 (aa381-483) expressed in E.coli.
Mouse monoclonal antibody raised against MLH1

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Tanyi et al.
European journal of surgical oncology : the journal of the European Society of Surgical Oncology and the British Association of Surgical Oncology, 40(11), 1445-1452 (2014-08-12)
Hereditary Non-Polyposis Colorectal Cancer is an inherited disease with deleterious germline mutations in the DNA mismatch repair genes causing the development of colon cancer and other malignancies. This is the first study in Hungary screening the population of our colorectal
Michael S Landau et al.
Human pathology, 45(8), 1704-1712 (2014-06-09)
Reduced CDX2 and cytokeratin 20 (CK20) expression in colorectal carcinoma with BRAF mutation and high-level microsatellite instability (MSI-H) has been well documented. The immunophenotype of BRAF-mutated microsatellite stable (MSS) colorectal carcinoma has not been reported. We analyzed 205 colorectal carcinomas
Raul Alvarado-Bachmann et al.
Molecular medicine reports, 10(2), 1003-1006 (2014-05-27)
Knowledge of the molecular biology of primary colorectal cancer (CRC) has improved in recent years, and one reason for this is the identification of microsatellite instability (MSI), which occurs in up to 15% of sporadic CRC. However, less is known
Koah R Vierkoetter et al.
Gynecologic oncology, 135(1), 81-84 (2014-08-06)
Patients with Lynch Syndrome are at an increased risk for a variety of malignancies, including ovarian cancer. Ovarian cancers associated with Lynch Syndrome are predominantly clear cell or endometrioid in histology. Lynch Syndrome is characterized by germline mutations in mismatch
Anne-Sophie Bats et al.
Anticancer research, 34(6), 3211-3215 (2014-06-13)
To assess the feasibility of Microsatellite Instability (MSI) analysis in uterine cavity washings for detecting endometrial cancer in Lynch syndrome. This was a proof-of-concept study in Lynch syndrome patients, scheduled for hysterectomy. At the beginning of surgical procedure, uterine cavity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service