Skip to Content
MilliporeSigma
All Photos(1)

Documents

SAB5300165

Sigma-Aldrich

Monoclonal Anti-OVA antibody produced in mouse

clone 3G2E1D9, ascites fluid

Synonym(s):

N/A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

3G2E1D9, monoclonal

technique(s)

direct ELISA: 1:10,000
western blot: 1:500-1:2,000

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

General description

Ovalbumin (323-339) is an egg white protein surrounded by B-cell epitopes, which are bound by specific IgE antibodies. In addition to this, it also contains CD4+ T cell epitopes.

Immunogen

Ovalbumin
Mouse monoclonal antibody raised against OVA

Biochem/physiol Actions

Ovalbumin (OVA) peptide 323-339 mediates 25-35% of T-cell response to intact OVA in isolated BALB/c mouse. OVA 323-339 has been widely used to determine the characteristics of class II major histocompatibility complex (MHC)-peptide binding and T-cell activation. It is extensively used in allergy research.

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Keigo Kondou et al.
Journal of biological engineering, 13, 77-77 (2019-10-28)
The baculovirus (BV) Autographa californica multiple nuclear polyhedrosis virus has been used in numerous protein expression systems because of its ability to infect insect cells and serves as a useful vaccination vector with several benefits, such as its low clinical
Ingrid S Zeelenberg et al.
Cancer research, 68(4), 1228-1235 (2008-02-19)
Expression of non-self antigens by tumors can induce activation of T cells in vivo, although this activation can lead to either immunity or tolerance. CD8+ T-cell activation can be direct (if the tumor expresses MHC class I molecules) or indirect
Comparison between ovalbumin and ovalbumin peptide 323-339 responses in allergic mice: humoral and cellular aspects.
Sun LZ
Scandinavian Journal of Immunology, 71(5), 329-335 (2010)
Tian-Yu Song et al.
Nature communications, 12(1), 7003-7003 (2021-12-03)
Cancer cells acquire genetic heterogeneity to escape from immune surveillance during tumor evolution, but a systematic approach to distinguish driver from passenger mutations is lacking. Here we investigate the impact of different immune pressure on tumor clonal dynamics and immune

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service