Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

SAB4504488

Sigma-Aldrich

Anti-phospho-NF-kappaB p65 (pSer276) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CMCU, Anti-NFKB3, Anti-p65

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 60 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer276)

Gene Information

human ... RELA(5970)

Immunogen

The antiserum was produced against synthesized peptide derived from human NF-kappaB p65 around the phosphorylation site of Ser276.

Immunogen Range: 249-298

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rie Hasebe et al.
The Journal of experimental medicine, 219(6) (2022-05-18)
Neural circuits between lesions are one mechanism through which local inflammation spreads to remote positions. Here, we show the inflammatory signal on one side of the joint is spread to the other side via sensory neuron-interneuron crosstalk, with ATP at
Kaoru Murakami et al.
Frontiers in immunology, 12, 675909-675909 (2021-06-12)
The lipopolysaccharides (LPSs) of Rhodobacter are reported to be TLR4 antagonists. Accordingly, the extract of Rhodobacter azotoformans (RAP99) is used as a health supplement for humans and animals in Japan to regulate immune responses in vivo. We previously analyzed the
Xiaozhi Deng et al.
Experimental and therapeutic medicine, 20(3), 2227-2235 (2020-08-09)
Inflammatory bowel disease (IBD) is a term used to describe chronic and recurrent gastrointestinal disease. In total, >2 million individuals worldwide have been diagnosed with IBD, including ulcerative colitis (UC), Crohn's disease (CD) and indeterminate colitis. There is accumulating evidence
Mitsutoshi Ota et al.
Arthritis & rheumatology (Hoboken, N.J.), 72(6), 931-942 (2019-12-01)
We have previously reported that the coactivation of NF-κB and STAT3 in nonimmune cells, including synovial fibroblasts, enhances the expression of NF-κB target genes and plays a role in chronic inflammation and rheumatoid arthritis (RA). This study was undertaken to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service