Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4503092

Sigma-Aldrich

Anti-XRCC3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

DNA-repair protein XRCC3, X-ray repair cross-complementing protein 3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 37 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... XRCC3(7517)

General description

Anti-XRCC3 Antibody detects endogenous levels of total XRCC3 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human XRCC3.

Immunogen Range: 41-90

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Hu et al.
PloS one, 8(8), e72104-e72104 (2013-08-27)
XRCC3 and RAD51 are two important members in homologous recombination repair pathway. This study was performed to detect the expressions of these two molecules in breast cancer and explore their correlations with clinicopathological factors. Immunohistochemistry was used to detect protein
Saudade André et al.
International journal of molecular sciences, 21(8) (2020-04-17)
Male breast cancer (BC) is a distinct neoplasm with low but rising incidence, frequently diagnosed as advanced stage disease. Considering the relevance of altered homologous recombination repair (HRR) in male BC, we aimed to explore the biomarker potential of aberrant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service