Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4502834

Sigma-Aldrich

Anti-SOX9 Antibody

rabbit polyclonal

Synonym(s):

SRY sex determining region Y-box 9 protein, SRY-box containing gene 9, Sox-9, transcription factor SOX-9

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
IF
IHC
WB
clone:
polyclonal
species reactivity:
human, mouse
citations:
5
technique(s):
ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

Product Name

Anti-SOX9 antibody produced in rabbit, affinity isolated antibody

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 56 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOX9(6662)

General description

Anti-SOX9 antibody detects endogenous levels of total SOX9 protein.
The SOX9 (SRY (sex determining region Y)-box 9) gene is located on human chromosome 17q24.3. SOX9 codes for a DNA-binding protein that belongs to the class of high-mobility-group box.

Immunogen

The antiserum was produced against synthesized peptide derived from human SOX9.

Immunogen Range: 147-196

Biochem/physiol Actions

The protein encoded by SOX9 (SRY (sex determining region Y)-box 9) gene is involved in the regulation of developmental processes such as chondrogenesis, gliogenesis, epithelial hair follicle stem cell specification, gonadogenesis, normal development of septum, lung, cardiac valve and pancreas. Abnormal regulation of SOX9 promotes tumorigenesis in different tissues. SOX9 induces the process of metastasis in cancer cells. Overexpression of the gene is observed in prostate, breast, and colorectal cancers.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression and Clinical Significance of SOX9 in Renal Cell Carcinoma, Bladder Cancer and Penile Cancer.
Wan YP
Oncology research and treatment, 40(1-2), 15-20 (2017)
Loss of DDRGK1 modulates SOX9 ubiquitination in spondyloepimetaphyseal dysplasia.
Egunsola AT
The Journal of Clinical Investigation, 127(4), 1475-1484 (2017)
Position effects due to chromosome breakpoints that map approximately 900 Kb upstream and approximately 1.3 Mb downstream of SOX9 in two patients with campomelic dysplasia.
Velagaleti GV
American Journal of Human Genetics, 76(4), 652-662 (2005)
Hyun Mi Kang et al.
Cell reports, 14(4), 861-871 (2016-01-19)
The kidney has a tremendous capacity to regenerate following injury, but factors that govern this response are still largely unknown. We isolated cells from mouse kidneys with high proliferative and multi-lineage differentiation capacity. These cells expressed a high level of
SOX9 is a proliferation and stem cell factor in hepatocellular carcinoma and possess widespread prognostic significance in different cancer types.
Richtig G
PLoS ONE, 12(11), 1-15 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service