Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4502258

Sigma-Aldrich

Anti-ATRX antibody produced in rabbit

affinity isolated antibody

Synonym(s):

RAD54L, Transcriptional regulator ATRX, X-linked helicase II, X-linked nuclear protein, XH2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 282 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATRX(546)

General description

Anti-ATRX Antibody detects endogenous levels of total ATRX protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human ATRX.

Immunogen Range: 111-160

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Epigenetic dysregulation and poorer prognosis in DAXX-deficient pancreatic neuroendocrine tumours.
Christodoulos P Pipinikas et al.
Endocrine-related cancer, 22(3), L13-L18 (2015-04-23)
Priscila Ramos-Ibeas et al.
Molecular reproduction and development, 86(8), 1033-1043 (2019-06-19)
Although telomere length (TL) shortens with age in most tissues, an age-related increase in length has been described in sperm through a mechanism that is not yet fully understood. Changes in TL with age in the same individual have not
Priscila Ramos-Ibeas et al.
Molecular reproduction and development, 86(8), 1033-1043 (2019-06-19)
Although telomere length (TL) shortens with age in most tissues, an age-related increase in length has been described in sperm through a mechanism that is not yet fully understood. Changes in TL with age in the same individual have not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service