Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4502043

Sigma-Aldrich

Anti-OPRD1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

δ-opioid receptor, δ-type opioid receptor, K56, MSL-2, OPRD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 40 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
western blot: 1:500-1:1000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OPRD(4985)

General description

Anti-OPRD1 antibody detects endogenous levels of total OPRD1 protein.
The OPRD1 (opioid receptor δ1) gene is mapped to human chromosome 1p35.3 and encodes for a G-protein-coupled receptor. It is abundantly expressed in peripheral tissues.

Immunogen

The antiserum was produced against synthesized peptide derived from human OPRD1.

Immunogen Range: 311-360

Biochem/physiol Actions

OPRD1 (opioid receptor δ1) is associated with cognitive function and mood driven behavior. Anxiolytic and antidepressant actions are brought about when specific agonists bind to the OPRD1 gene. δ-opioid receptor (DOR) is known to be involved in the progression and development of specific tumours (malignant). Upregulation of this gene is observed in breast cancer, and thus, it can serve as an important therapeutic marker.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Louis Gendron et al.
Pharmacological reviews, 68(3), 631-700 (2016-06-28)
Opioids are among the most effective analgesics available and are the first choice in the treatment of acute severe pain. However, partial efficacy, a tendency to produce tolerance, and a host of ill-tolerated side effects make clinically available opioids less
Ann M Manzardo et al.
Gene, 560(2), 184-194 (2015-02-07)
Alcoholism arises from combined effects of multiple biological factors including genetic and non-genetic causes with gene/environmental interaction. Intensive research and advanced genetic technology has generated a long list of genes and biomarkers involved in alcoholism neuropathology. These markers reflect complex
Yang-Chao Wei et al.
Oncology reports, 36(5), 2579-2586 (2016-10-26)
δ‑opioid receptor (DOR) belongs to the family of G protein‑coupled receptors (GPCRs). Numerous studies have shown that DOR is widely distributed in human peripheral tissues and is closely related to the development and progression of certain malignant tumours. However, there is controversy in
Huihui Ji et al.
PloS one, 12(3), e0172335-e0172335 (2017-03-03)
Aberrant DNA methylation has been observed in the patients with Alzheimer's disease (AD), a common neurodegenerative disorder in the elderly. OPRD1 encodes the delta opioid receptor, a member of the opioid family of G-protein-coupled receptors. In the current study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service