Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4501764

Sigma-Aldrich

Anti-Lamin A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

70 kDa lamin, LAMA, LMN1, LMNA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 74 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LMNA(4000)

General description

Lamin A/C (LMNA) gene codes for lamin A and C. Lamin A/C is a nuclear intermediate filament protein. LMNA gene is mapped to human chromosome 1q22. Anti-Lamin A Antibody detects endogenous levels of total Lamin A protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Lamin A/C.

Immunogen Range: 361-410

Application

Anti-Lamin A antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Lamin A/C (LMNA) is involved in the structural organization of the nucleus. It modulates gene expression. It may play a role in osteoblastogenesis. Mutation in the LMNA gene is associated with muscle dystrophies, cardiomyopathies, lipodystrophies, and neuropathies. LMNA protein plays a vital role in cell cycle progression. It controls extracellular signal-regulated kinase (ERK)/mitogen-activated protein kinase (MAPK) and Wnt β-catenin pathway.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Sowińska-Seidler et al.
Journal of applied genetics, 59(3), 281-289 (2018-05-31)
Craniosynostosis (CS) refers to the group of craniofacial malformations characterized by the premature closure of one or more cranial sutures. The disorder is clinically and genetically heterogeneous and occurs usually as an isolated trait, but can also be syndromic. In
Halida P Widyastuti et al.
BMC medical genetics, 21(1), 152-152 (2020-07-24)
Intermediate filament proteins that construct the nuclear lamina of a cell include the Lamin A/C proteins encoded by the LMNA gene, and are implicated in fundamental processes such as nuclear structure, gene expression, and signal transduction. LMNA mutations predominantly affect
Hin Chu et al.
The Journal of biological chemistry, 293(30), 11709-11726 (2018-06-12)
Coronavirus tropism is predominantly determined by the interaction between coronavirus spikes and the host receptors. In this regard, coronaviruses have evolved a complicated receptor-recognition system through their spike proteins. Spikes from highly related coronaviruses can recognize distinct receptors, whereas spikes
Elena V Ignatieva et al.
Genes, 11(9) (2020-09-11)
Laminopathies are a family of monogenic multi-system diseases resulting from mutations in the LMNA gene which include a wide range of neuromuscular disorders. Although lamins are expressed in most types of differentiated cells, LMNA mutations selectively affect only specific tissues
Lorenzo Bazzani et al.
Oncotarget, 9(19), 14939-14958 (2018-03-31)
Prostaglandin E2 (PGE2) contributes to tumor progression by promoting cancer cell growth, invasion and by creating a favorable pro-tumor microenvironment. PGE2 has been reported to transactivate and internalize into the nucleus receptor tyrosine kinases such as Epidermal growth factor receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service