Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4500888

Sigma-Aldrich

Anti-FGFR3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CD333, CD333 antigen, FGFR-3, Fibroblast growth factor receptor 3, JTK4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 87 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FGFR3(2261)

General description

Anti-FGFR3 Antibody detects endogenous levels of total FGFR3 protein.

FGFR3 (fibroblast growth factor receptor 3) belongs to the fibroblast growth factor receptor (Fgfr) family of proteins. It has 19 exons, an extracellular ligand binding domain, a transmembrane domain and an intracellular tyrosine kinase domain. It is located on human chromosome 4p16.3.

Immunogen

The antiserum was produced against synthesized peptide derived from human FGFR3.

Immunogen Range: 131-180

Application

Anti-FGFR3 antibody has been used in immunohistochemistry.

Biochem/physiol Actions

Germline mutations in FGFR3 (fibroblast growth factor receptor 3) results in autosomal dominant skeletal dysplasias like achondroplasia (ACH), thanatophoric dysplasia (TD) and hypochrondroplasia (HCH). The gene also controls cellular activity.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loss of heterozygosity at 4p16.3 and mutation of FGFR3 in transitional cell carcinoma
Sibley K, et al.
Oncogene, 20, 686-691 (2001)
Masanobu Kawai et al.
The Journal of biological chemistry, 288(4), 2414-2427 (2012-12-14)
Fibroblast growth factor-23 (FGF23) is well established to play crucial roles in the regulation of phosphate homeostasis. X-linked hypophosphatemic rickets (XLH) is characterized by impaired mineralization and growth retardation associated with elevated circulating FGF23 levels. Administration of phosphate and calcitriol
Daniella Da Silva et al.
Regenerative therapy, 24, 602-616 (2023-11-30)
Worldwide, osteoporosis is the utmost predominant degenerative bone condition. Stem cell regenerative therapy using adipose-derived mesenchymal stem cells (ADMSCs) is a promising therapeutic route for osteoporosis. Photobiomodulation (PBM) has sparked considerable international appeal due to its' ability to augment stem

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service