Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4500249

Sigma-Aldrich

Anti-PARK7 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Oncogene DJ1, Parkinson disease protein 7, Protein DJ-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 19 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PARK7(11315)

General description

Anti-PARK7 Antibody detects endogenous levels of total PARK7 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human PARK7.

Immunogen Range: 61-110

Application

Anti-PARK7 antibody produced in rabbit is suitable for indirect ELISA and western blot applications.

Biochem/physiol Actions

PARK7 is a member of novel glyoxalase family. It exerts mitochondrial activity. It has been suggested that PARK7 may act as an antioxidant and/or a molecular chaperone. It may also detoxify aggressive metabolic aldehyde by-products. It helps in stabilizing mitochondria during cellular stress. Alteration in gene function affects neurodegeneration, which leads to the autosomal recessive early-onset parkinsonism.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacek R Wiśniewski et al.
Journal of proteome research, 15(7), 2321-2326 (2016-06-15)
Proteomics and other protein-based analysis methods such as Western blotting all face the challenge of discriminating changes in the levels of proteins of interest from inadvertent changes in the amount loaded for analysis. Mass-spectrometry-based proteomics can now estimate the relative
Yusuke Toyoda et al.
Biology open, 3(8), 777-784 (2014-07-27)
Parkinson's disease is associated with mitochondrial decline in dopaminergic neurons of the substantia nigra. One of the genes linked with the onset of Parkinson's disease, DJ-1/PARK7, belongs to a novel glyoxalase family and influences mitochondrial activity. It has been assumed
V Bonifati et al.
Neurological sciences : official journal of the Italian Neurological Society and of the Italian Society of Clinical Neurophysiology, 24(3), 159-160 (2003-11-05)
Four chromosomal loci ( PARK2, PARK6, PARK7, and PARK9) associated with autosomal recessive, early onset parkinsonism are known. We mapped the PARK7 locus to chromosome 1p36 in a large family from a genetically isolated population in the Netherlands, and confirmed
Vincenzo Bonifati et al.
Science (New York, N.Y.), 299(5604), 256-259 (2002-11-26)
The DJ-1 gene encodes a ubiquitous, highly conserved protein. Here, we show that DJ-1 mutations are associated with PARK7, a monogenic form of human parkinsonism. The function of the DJ-1 protein remains unknown, but evidence suggests its involvement in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service