Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4301182

Sigma-Aldrich

Anti-MUC6 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

MUC-6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

concentration

1.5 mg/mL

technique(s)

immunohistochemistry: 1:50- 1:200

isotype

IgG

accession no.

NP_005952.2

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUC6(4588)

Specificity

The antibody detects endogenous levels of total MUC6 protein.

Immunogen

Synthetic peptide corresponding to a region derived from internal residues of human mucin 6, oligomeric mucus/gel-forming

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.3 PBS, 0.05% NaN3, 50% Glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Kyu Lee et al.
Pathology, research and practice, 215(3), 459-465 (2018-12-26)
ALK-positive (ALK+) lung adenocarcinoma usually shows a more advanced-staged disease with frequent nodal metastasis and highly aggressive outcomes compared with EGFR-mutated lung cancers. The aim of this study was to investigate the expression profiles of several mucins in ALK + lung cancers
Jingsun Wei et al.
Cancer management and research, 11, 1275-1287 (2019-02-26)
Gastric hepatoid adenocarcinoma is a rare subtype of primary gastric cancer and is a high-grade form of malignancy. However, the pathogenesis and molecular biology of gastric hepatoid adenocarcinoma remain poorly understood. The aim of this study was to establish and
Purushothama Rao Tata et al.
Developmental cell, 44(6), 679-693 (2018-03-28)
We show that the loss or gain of transcription factor programs that govern embryonic cell-fate specification is associated with a form of tumor plasticity characterized by the acquisition of alternative cell fates normally characteristic of adjacent organs. In human non-small

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service