Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4301113

Sigma-Aldrich

Anti-HRAS Antibody

rabbit polyclonal

Synonym(s):

CTLO, HAMSV, HRAS1, K-RAS, v-Ha-ras Harvey rat sarcoma viral oncogene homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-RAS antibody produced in rabbit, affinity isolated antibody

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

21 kDa

species reactivity

mouse, rat, human

concentration

0.7 mg/mL

technique(s)

western blot: 1:200-1:1000 (Cell Lysate)

isotype

IgG

accession no.

NP_001123914.1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HRAS(3265)

General description

Ras is a proto-oncogene and the gene encoding it is localized on human chromosome 11p15.5. It is a 21 kDa protein and is part of the small GTP-binding proteins superfamily.

Specificity

The antibody detects endogenous level of total RAS protein.

Immunogen

Synthetic peptide corresponding to a region derived from 60-74 amino acids of Human v-Ha-ras Harvey rat sarcoma viral oncogene homolog

Biochem/physiol Actions

Ras is a guanine-nucleotide binding protein that couples tyrosine kinase receptor signals to mitogen activated protein (MAP) kinase cascade. It has an important role in cell growth. Ras is a substrate for farnesyltransferase (FTase) and when farnesylated, it is anchored to the membrane. Membrane localization of Ras is crucial for its cellular activity. Inhibition of Ras farnesylation is a target for anti-cancer drug development.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.7mg/mL in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3, 0.05% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human mucin gene MUC5B, the 10.7-kb large central exon encodes various alternate subdomains resulting in a super-repeat structural evidence for a 11p15. 5 gene family.
Desseyn J L, et al.
The Journal of Biological Chemistry, 272(6), 3168-3178 (1997)
Germline mutations in HRAS proto-oncogene cause Costello syndrome.
Aoki Y, et al.
Nature Genetics, 37(10), 1038-1038 (2005)
Ras protein farnesyltransferase: A strategic target for anticancer therapeutic development.
Rowinsky E K, et al.
Journal of Clinical Oncology, 7(11), 3631-3652 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service