Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB4301078

Sigma-Aldrich

Anti-TDP2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

AD022, EAP2, EAPII, TTRAP, hTDP2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

41 kDa

species reactivity

human, mouse

concentration

2.6 mg/mL

technique(s)

immunohistochemistry: 1:50- 1:200
western blot: 1:200-1:1000 (Cell Lysate)

isotype

IgG

accession no.

NP_057698.2

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TDP2(51567)

General description

Tyrosyl DNA phosphodiesterase-2 (TDP2) is a multi-functional protein and is also called as TTRAP (TNF receptor-associated protein). It is a 5- tyrosyl DNA phosphodiesterase that belongs to the PML NB (promyelocytic leukemia nuclear body) protein family. TDP2 is mapped to human chromosome 6p22.

Specificity

The antibody detects endogenous levels of total TDP2 protein.

Immunogen

Synthetic peptide corresponding to a region derived from internal residues of human tyrosyl-DNA phosphodiesterase 2

Biochem/physiol Actions

Chromosomal translocations stimulated by DNA topoisomerase II at the time of gene transcription is repressed by tyrosyl DNA phosphodiesterase-2 (TDP2). It is essential to preserve the chromosome stability in human cells. TDP2 participates in cell proliferation, apoptosis and the repair of DNA damage. It promotes cell cycle arrest and apoptosis in osteosarcoma cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.3 PBS, 0.05% NaN3, 50% Glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tyrosyl-DNA-phosphodiesterases (tdp1 and tdp2).
Pommier Y, et al.
DNA Repair, 19, 114-129 (2014)
Overexpression of TTRAP inhibits cell growth and induces apoptosis in osteosarcoma cells.
Zhou C, et al.
Bmb Reports, 46(2), 113-113 (2013)
TDP2 suppresses chromosomal translocations induced by DNA topoisomerase II during gene transcription.
Gomez-Herreros F, et al.
Nature Communications, 8(1), 233-233 (2017)
Henrike Johanna Gothe et al.
Molecular cell, 75(2), 267-283 (2019-06-17)
How spatial chromosome organization influences genome integrity is still poorly understood. Here, we show that DNA double-strand breaks (DSBs) mediated by topoisomerase 2 (TOP2) activities are enriched at chromatin loop anchors with high transcriptional activity. Recurrent DSBs occur at CCCTC-binding factor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service