Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4300623

Sigma-Aldrich

Anti-Tubulin β-III antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CFEOM3A, Anti-TUBB3, Anti-TUBB4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~55 kDa

species reactivity

mouse, human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(E-E-S-E-A)

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TUBB3(10381)

General description

The antibody detects endogenous level of total Tubulin -βIII protein.
Tubulin β-III (TUBB3) is a constituent of microtubules. It is expressed in mature neurons. It is located on human chromosome 16q24.

Immunogen

Peptide sequence around aa. 442-446 (E-E-S-E-A), according to the protein NP_006077.2

Application

Anti-Tubulin β-III antibody has been used in immunocytochemistry and western blot analysis.

Biochem/physiol Actions

Upregulation of tubulin β-III (TUBB3) shows vigorous tumor characteristics and genetic instability in urinary bladder cancer. It participates in the separation of chromosome. Individuals with congenital fibrosis of the extraocular muscle type 3 (CFEOM3) show mutations in TUBB3. βIII-tubulin modulates glucose metabolism, thereby transforms glucose starvation stress signaling to develop the multiplication of cell and survival in non-small cell lung cancer cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha-chain. TUBB3 plays a critical role in proper axon guidance and mantainance.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J T Corthell et al.
Neuroscience, 222, 38-48 (2012-07-21)
Circadian rhythms affect olfaction by an unknown molecular mechanism. Independent of the suprachiasmatic nuclei, the mammalian olfactory bulb (OB) has recently been identified as a circadian oscillator. The electrical activity in the OB was reported to be synchronized to a
Genome-wide gene amplification during differentiation of neural progenitor cells in vitro
Fischer U, et al.
PLoS ONE, 7(5) (2012)
Mutations in Human Tubulin Proximal to the Kinesin-Binding Site Alter Dynamic Instability at Microtubule Plus- and Minus-Ends
Ti SC, et al.
Developmental Cell, 37(1), 72-84 (2016)
Directed differentiation of postnatal hippocampal neural stem cells generates nuclear receptor related-1 protein- and tyrosine hydroxylase-expressing cells
Ding Y, et al.
Molecular Medicine Reports, 14(3), 1993-1999 (2016)
?III-tubulin overexpression is linked to aggressive tumor features and genetic instability in urinary bladder cancer
Hinsch A, et al.
Human Pathology, 61, 210-220 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service