Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200823

Sigma-Aldrich

Anti- Protease-7 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Omptin, Outer membrane protein 3B, Protease A, Protease VII

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

antibody form

affinity isolated antibody

antibody product type

primary antibodies

description

Research area: Microbiome

form

buffered aqueous solution

species reactivity

E. coli, Shigella flexneri

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunoblotting: 0.5-1 μg/mL using E. coli lysate.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Escherichia coli K12 ... ompT(945185)
Shigella flexneri ... ompT(1238162)

Related Categories

General description

E. coli Protease-7, also known as Omptin (ompT) or Outer membrane protein 3B, belongs to the omptin family of outer membrane proteases found in several Gram-negative bacteria of the Enterobacteriaceae family. Omptins are unique proteases combining features of both serine and aspartate proteases activity. Several group members may also have cysteine protease and metalloproteases activities, all share a conserved active site. The enzyme proteolytic activity is dependent on the interaction with the lipid A region of the lipopolysaccharide (LPS).

Immunogen

Synthetic peptide corresponding to human N-terminal region E.coli Protease7, conjugated to KLH

Application

Anti-Protease-7 antibody specifically recognizes Protease-7 from E. coli and Shigella flexneri and does not cross react with Proteus mirabilis. The antibody may be used in various immunochemical techniques including Immunoblotting (doublet ~35 kDa).

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Disclaimer

This product is for R&D use only, not for drug, household, or other uses.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service