Skip to Content
MilliporeSigma
All Photos(1)

Documents

SAB4200735

Sigma-Aldrich

Anti-CRISPR/CAS9 -FITC antibody, Mouse monoclonal

clone 7A9-3A3, purified from hybridoma cell culture

Synonym(s):

Anti-CRISPR-associated protein-9 nuclease, Anti-Crispr, Anti-Crispr RNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

conjugate

FITC conjugate

Quality Level

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

7A9-3A3, monoclonal

form

buffered aqueous solution

storage condition

protect from light

concentration

~1.0 mg/mL

technique(s)

immunofluorescence: 1:200-1:400 using human HEK-293T cells over-expressing CAS9 protein

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats) belongs to the type II CRISPR/CAS9 system. It is part of an adaptive immune system of the Streptococcus pyogenes SF370, protecting from pathogens′ target genes by cleaving the foreign DNA in a sequence-dependent manner.
The type II CRISPR/Cas system has been adapted to expression in eukaryotic cells. The Cas9 endonuclease can be engineered with a single gRNA (guide RNA), directing a DNA double-strand break (DSB) at a desired genomic location. As a result, the cell activates endogenous DNA repair processes, either non-homologous end joining (NHEJ) or homology-directed repair (HDR). In comparison to other genome-editing technologies such as designer zinc fingers (ZFs), transcription activator–like effectors (TALEs) and homing meganucleases, the CRISPR/CAS9 system is a scalable, affordable and easy to engineer. Therefore, the anti-CRISPR/CAS9-FITC conjugated antibody can be a useful tool for detecting CRISPR/CAS9 positively transfected cells, revealing DSB sites in their genome.

Immunogen

Recombinant protein within the N-terminal region of Streptococcus pyogene Cas9

Application

Monoclonal Anti-CRISPR/CAS9-FITC recognizes the CAS9 protein in CAS9 construct over-expression systems. The antibody may be used in various immunochemical techniques including Immunofluorescence. The antibody successfully recognizes mutant Cas9 variants, including nickase Cas9 and dead Cas9 (dCas9) and does not cross react with FnCas9 from Francisella novicida bacteria and Cpf1 proteins from Acidaminococcus sp. (strain BV3L6) and Lachnospiraceae bacterium ND2006.

Physical form

Solution in 0.01M phosphate buffered saline pH 7.4 containing 1% bovine serum albumin and 15mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C, protected from light for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Other Notes

This product is for R&D use only, not for drug, household, or other uses.
In order to obtain best results in different techniques and preparations we recommend determining optimal working concentration by titration test.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service