Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200734

Sigma-Aldrich

Anti-IL6 high affinity antibody, Mouse monoclonal

clone IL6-2A3, purified from hybridoma cell culture

Synonym(s):

Anti-B-cell stimulatory factor 2 (BSF-2), Anti-CTL differentiation factor (CDF), Anti-Hybridoma growth factor, Anti-Interferon beta-2 (IFN-beta-2), Anti-Interleukin-6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

IL6-2A3, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

ELISA: suitable
immunoblotting: 0.125-0.25 μg/mL using recombinant human IL6 expressed in HEK-293T cells
immunoprecipitation (IP): 10-20 μg/test using concentrated supernatant of human osteosarcoma MG-63 cells treated with IL1β

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL6(3569)

General description

Interleukin-6 (IL6) is a multifunctional cytokine.

Immunogen

recombinant human IL6

Application

Anti-IL6 high affinity antibody, Mouse monoclonal has been used in
  • immunoblotting
  • immunoprecipitation
  • ELISA

Biochem/physiol Actions

Interleukin-6 (IL6) plays a wide variety of important biological roles in the immune, nervous and endocrine system. It is an essential component for hybridoma growth and is found in many supplemental cloning media. IL6 regulates acute and chronic inflammatory diseases and has been proven to be an excellent target for clinical treatment against autoimmune disorders such as rheumatoid arthritis, juvenile idiopathic arthritis and Castleman′s disease. IL6 also plays a key role in the pathogenesis of multiple myeloma.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IL-6 in autoimmune disease and chronic inflammatory proliferative disease
Ishihara K and Hirano T
Cytokine & Growth Factor Reviews, 13(4-5), 357-368 (2002)
A phase 2 multicentre study of siltuximab, an anti-interleukin-6 monoclonal antibody, in patients with relapsed or refractory multiple myeloma
Voorhees PM, et al.
British Journal of Haematology, 161(3), 357-366 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service