Skip to Content
MilliporeSigma
All Photos(3)

Documents

SAB4200399

Sigma-Aldrich

Anti-WIPI-2 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-ATG18B, Anti-Atg21, Anti-CGI-50, Anti-WIPI2, WD repeat domain, phosphoinositide interacting 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~49 kDa

species reactivity

rat, mouse, human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 2.5-5.0 μg using lysates of rat NRK cells.
western blot: 1-2 μg/mL using using whole extracts of HEK-293T cells over-expressing mouse WIPI-2.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WIPI2(26100)
mouse ... Wipi2(74781)
rat ... Wipi2(288498)

General description

The WIPI2 gene is mapped on the human chromosome at 7p22.1. WIPI2 has a 7-bladed propeller structure and contains a conserved motif.
WD repeat domain, phosphoinositide-interacting 2 (WIPI-2) belongs to the WIPI protein family. It is localized on the autophagy membrane and plasma membrane. Apart from this, it is also present on the membranes near the Golgi cisternae. WIPI2 is a mammalian ortholog of Atg18, which is ubiquitously expressed in a variety of cell lines.

Specificity

Anti-WIPI-2 recognizes human, rat, and mouse WIPI-2.

Application

Anti-WIPI-2 antibody produced in rabbit may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

WIPI-2 (WD repeat domain, phosphoinositide-interacting 2) plays a vital role in the transformation of omegasomes into autophagosomes. The above protein functions as a mammalian effector of phosphatidylinositol 3-phosphate (PtdIns3P). WIPI2 aids recombinant capsid protein VP1 (rVP1) in upregulating autophagy, MAPK1 (Mitogen-activated protein kinase 1)/3 phosphorylation and matrix metallopeptidase 9 (MMP-9) activity, which are required for macrophage migration.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mammalian Atg18 (WIPI2) localizes to omegasome-anchored phagophores and positively regulates LC3 lipidation.
Polson HE
Autophagy (2010)
Recombinant protein rVP1 upregulates BECN1-independent autophagy, MAPK1/3 phosphorylation and MMP9 activity via WIPI1/WIPI2 to promote macrophage migration.
Liao CC
Autophagy (2013)
Melanie Haas Kucherlapati
Oncotarget, 10(65), 6913-6933 (2019-12-21)
Genes of the pre-replication, pre-initiation and replisome complexes duplicate the genome from many sites once in a normal cell cycle. This study examines complex components in lung adenocarcinoma (LUAD) closely, correlating changes in the genome and transcriptome with proliferation and
Freeze-fracture replica immunolabelling reveals human WIPI-1 and WIPI-2 as membrane proteins of autophagosomes.
Proikas-Cezanne T
Journal of Cellular and Molecular Medicine (2011)
Wei Wan et al.
Molecular cell, 72(2), 303-315 (2018-10-20)
mTORC1, the major homeostatic sensor and responder, regulates cell catabolism mainly by targeting autophagy. Here, we show that mTORC1 directly controls autophagosome formation via phosphorylation of WIPI2, a critical protein in isolation membrane growth and elongation. mTORC1 phosphorylates Ser395 of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service