Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200321

Sigma-Aldrich

Anti-PTPRF (C-terminal) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-LAR, Anti-Leukocyte antigen-related tyrosine phosphatase, Anti-Leukocyte common antigen-related tyrosine phosphatase, Anti-Tyrosine protein phosphatase receptor type F

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~150 kDa
antigen ~210 kDa
antigen ~85 kDa

species reactivity

human

technique(s)

immunoprecipitation (IP): 5-10 μL using lysates of HEK-293T cells over-expressing human PTPRF.
western blot: 1:500-1:1000 using lysates of HEK-293T cells over-expressing human PTPRF.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTPRF(5792)
mouse ... Ptprf(19268)
rat ... Ptprf(360406)

Related Categories

General description

PTPRF or leukocyte common antigen-related protein (LAR) is a widely expressed protein tyrosine phosphatase with an extracellular receptor region that resembles a cell adhesion molecule.
Protein tyrosine phosphatase, receptor type F (PTPRF) consists of noncovalently bound subunits designated as E (extracellular) and P (phosphatase) subunits that are derived from a precursor protein by proteolytic processing. The E subunit (150 kDa) has three immunoglobulin-like domains and eight fibronectin III-like domains, and the P subunit (85 kDa) has a short extracellular domain, a transmembrane segment and tandem cytoplasmic PTP catalytic domains.

Specificity

The antibody recognizes both the extracellular and the cytoplasmic subunits of human PTPRF. Anti-PTPRF (C-terminal) antibody is specific for the P- subunit and the whole PTPRF protein from Human origin. Detection of the PTPRF bands by immunoblotting is specifically inhibited by the immunizing peptide.

Application

Anti-PTPRF (C-terminal) antibody produced in rabbit has been used in an enzyme based assay of protein tyrosine phosphatase 1 (PTP1).
The antibody may be used in several immunochemical techniques including immunoblotting (∼210, ∼150, and ∼85 kDa) and immunoprecipitation.

Biochem/physiol Actions

Protein tyrosine phosphatase, receptor type F (PTPRF) removes phosphate group from β-catenin, an event that may subsequently facilitate cell-cell adhesion and ensure the stability of the cadherin complex. This phosphatase has also been implicated in various cellular processes such as neurite growth, nerve regeneration, actin remodeling and regulation of insulin function. Insulin receptor, epidermal growth factor receptor (EGF) receptor, hepatocyte growth factor (HGF) receptor and rearranged during transfection (RET) have been reported as substrates of PTPRF.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differential effects of leukocyte common antigen-related protein on biochemical and biological activities of RET-MEN2A and RET-MEN2B mutant proteins
Qiao S, et al.
The Journal of Biological Chemistry, 276(12), 9460-9467 (2001)
2, 5-DISUBSTITUTED OXADIAZOLE AND IT?S DERIVATIVE INHIBIT PROTEIN TYROSINE PHOSPHATASE PTP1B, ENHANCING INSULIN RECEPTOR PHOSPHORYLATION
Athare CL, et al.
World Journal of Pharmaceutical Research, 3(2), 2232-2247 (2014)
Mélanie J Chagnon et al.
Biochemistry and cell biology = Biochimie et biologie cellulaire, 82(6), 664-675 (2005-01-28)
The protein tyrosine phosphatases (PTPs) have emerged as critical players in diverse cellular functions. The focus of this review is the leukocyte common antigen-related (LAR) subfamily of receptor PTPs (RPTPs). This subfamily is composed of three vertebrate homologs, LAR, RPTP-sigma
Robert A Mooney et al.
Current topics in medicinal chemistry, 3(7), 809-819 (2003-04-08)
The leukocyte common antigen-related protein, LAR, is a receptor-like protein tyrosine phosphatase (PTP) which has a wide tissue distribution. Post-translational processing cleaves the proprotein into two non-covalently associated subunits, an extracellular subunit resembling a cell adhesion molecule with three immunoglobulin-like
William J Polacheck et al.
Nature, 552(7684), 258-262 (2017-11-22)
The vascular barrier that separates blood from tissues is actively regulated by the endothelium and is essential for transport, inflammation, and haemostasis. Haemodynamic shear stress plays a critical role in maintaining endothelial barrier function, but how this occurs remains unknown.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service