Skip to Content
MilliporeSigma
All Photos(3)

Documents

SAB3500991

Sigma-Aldrich

Anti-CIITA antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 124 kDa

species reactivity

rat, mouse, human

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunoblotting: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CIITA(4261)

Immunogen

Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human CIITA.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500991.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Raval et al.
Molecular cell, 7(1), 105-115 (2001-02-15)
The CIITA coactivator is essential for transcriptional activation of MHC class II genes and mediates enhanced MHC class I transcription. We now report that CIITA contains an intrinsic acetyltransferase (AT) activity that maps to a region within the N-terminal segment
Inohara et al.
Annual review of biochemistry, 74, 355-383 (2005-06-15)
Nods are cytosolic proteins that contain a nucleotide-binding oligomerization domain (NOD). These proteins include key regulators of apoptosis and pathogen resistance in mammals and plants. A large number of Nods contain leucine-rich repeats (LRRs), hence referred to as NOD-LRR proteins.
J A Harton et al.
Science (New York, N.Y.), 285(5432), 1402-1405 (1999-08-28)
Class II transactivator (CIITA) is a global transcriptional coactivator of human leukocyte antigen-D (HLA-D) genes. CIITA contains motifs similar to guanosine triphosphate (GTP)-binding proteins. This report shows that CIITA binds GTP, and mutations in these motifs decrease its GTP-binding and
Andreas Neerincx et al.
Journal of immunology (Baltimore, Md. : 1950), 193(6), 3090-3100 (2014-08-17)
Ag presentation to CD4(+) and CD8(+) T cells depends on MHC class II and MHC class I molecules, respectively. One important regulatory factor of this process is the transcriptional regulation of MHC gene expression. It is well established that MHC
Salomé LeibundGut-Landmann et al.
European journal of immunology, 34(6), 1513-1525 (2004-05-27)
The class II transactivator (CIITA) has been referred to as the "master control factor" for the expression of MHC class II (MHCII) genes. As our knowledge on the specificity and function of CIITA grows, it is becoming increasingly evident that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service