Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB3500533

Sigma-Aldrich

Anti-AFP antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Alpha-fetoprotein, Anti-FETA, Anti-HPAFP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AFP(174)

General description

The gene AFP (α fetoprotein) encodes a plasma glycoprotein that has a molar mass of 70kDa. The gene is mapped to human chromosome 4. The gene is evolutionarily-related to serum albumin, which is also mapped to human chromosome 4. It is synthesized along with serum albumin during the development of embryonic yolk sac and the liver.

Immunogen

AFP antibody was raised against a 14 amino acid peptide near the amino terminus of human AFP.

Application

Anti-AFP antibody produced in rabbit has been used in immunoflorescence.

Biochem/physiol Actions

α-fetoprotein encoded by the gene AFP is involved in the regulation of fatty acids in both fetal and proliferating adult liver cells. Its expression is found to be increased in acute liver injuries, indicating active liver regeneration. It has been associated with fatty liver disease (FLD), a disease that may lead to cirrhosis and hepatocellular carcinoma. It serves as a tumor marker for HCC (hepatocellular carcinoma).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500533.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Samuel Fagg et al.
Cell transplantation, 30, 963689721993780-963689721993780 (2021-03-05)
Stem cell transplantation to the liver is a promising therapeutic strategy for a variety of disorders. Hepatocyte transplantation has short-term efficacy but can be problematic due to portal hypertension, inflammation, and sinusoidal thrombosis. We have previously transplanted small mouse endoderm
Mingna Zhang et al.
Cancer medicine, 12(17), 18032-18049 (2023-08-14)
In liver cancer, leucine-rich repeat-containing G-protein coupled receptor 5 (LGR5) compartment represents an important tumor-initiating cell (TIC) population and served as a potential therapeutic target. Cancer-associated fibroblasts (CAFs) is a critical part of the tumor microenvironment, heavily influenced TIC function
Wenwen Deng et al.
Journal of agricultural and food chemistry, 64(6), 1264-1273 (2016-01-27)
Pleurotus eryngii is a medicinal/edible mushroom with great nutritional value and bioactivity. Its polysaccharide has recently been developed into an effective gene vector via cationic modification. In the present study, cationized P. eryngii polysaccharide (CPS), hybridized with calcium phosphate (CP)
M E Harper et al.
American journal of human genetics, 35(4), 565-572 (1983-07-01)
Albumin and alpha-fetoprotein are structurally related serum proteins, having a similar gene structure and, conceivably, a common evolutionary origin. To test their relative arrangement in the human genome, the serum albumin and alpha-fetoprotein genes were mapped by in situ hybridization
Surachate Siripongsakun et al.
Journal of gastroenterology and hepatology, 29(1), 157-164 (2013-12-21)
The performance of alpha-fetoprotein (AFP) in the detection of hepatocellular carcinoma (HCC) recurrence after radiofrequency ablation was analyzed. One hundred and forty-six solitary HCC lesions treated by radiofrequency ablation were evaluated. Using the AFP cutoff level at ≥ 20 ng/mL, tumors were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service