Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB3500428

Sigma-Aldrich

Anti-DR4 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-TRAIL-R1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 56 kDa

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Soluble tumor necrosis factor-related apoptosis-inducing ligand-receptor 1 (sTRAIL-R1), also known as DR4, is encoded by the gene mapped to human chromosome 8p21-22. It belongs to the TNFR superfamily of transmembrane proteins. TRAIL-R1 contains a cytoplasmic "death domain," which can activate the cell′s apoptotic machinery. TRAIL receptor-1/DR4 is activated by binding to either membrane anchored or soluble TRAIL/Apo2L.

Immunogen

DR4 antibody was raised against a peptide corresponding to amino acids near the carboxy terminus of human DR4 protein.

Application

Anti-DR4 antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Soluble tumor necrosis factor-related apoptosis-inducing ligand-receptor 1 (sTRAIL-R1) interacts with tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) and stimulates cell-death signaling pathway. Mutation of the sTRAIL-R1 gene is associated with the pathogenesis of non-Hodgkin′s lymphoma (NHL). In addition, variation of TRAIL-R1 also increases the risk of susceptibility to prostate cancer (PCa) in North Indian population.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500428.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of TRADD in death receptor signaling.
Pobezinskaya YL and Liu Z
Cell Cycle (2012)
DDIT3 and KAT2A Proteins Regulate TNFRSF10A and TNFRSF10B Expression in Endoplasmic Reticulum Stress-mediated Apoptosis in Human Lung Cancer Cells*
Tianliang Li
The Journal of Biological Chemistry (2015)
Death receptor 4 variants enhanced prostate cancer risk in North Indian population.
Mittal RD
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine (2015)
Somatic mutations of TRAIL-receptor 1 and TRAIL-receptor 2 genes in non-Hodgkin's lymphoma.
Lee SH
Oncogene (2001)
Tianliang Li et al.
The Journal of biological chemistry, 290(17), 11108-11118 (2015-03-15)
TNFRSF10A and TNFRSF10B are cell surface receptors that bind to tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) and mediate the extrinsic pathway of apoptosis. However, the mechanisms of transcriptional regulation of TNFRSF10A and TNFRSF10B remain largely uncharacterized. In this study, two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service