Skip to Content
MilliporeSigma
All Photos(3)

Documents

SAB3500252

Sigma-Aldrich

Anti-TIM-1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-T cell immunoglobulin and mucin domain containing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 40 kDa

species reactivity

human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HAVCR1(26762)

General description

Hepatitis A virus cellular receptor 1 (HAVCR1) or T-cell immunoglobulin and mucin protein (TIM-1) is a type 1 transmembrane glycoprotein. The protein is found in the apical membrane of cells. It is expressed in the kidney, testis and activated CD4+ T-cells. It possesses a six-cysteine immunoglobulin-like domain, a T/SP rich domain and two N-glycosylation sites. The TIM-1 gene is localized on human chromosome 5q33.2.

Immunogen

TIM-1 antibody was raised against a 16 amino acid peptide from near the amino terminus of human TIM-1.

Application

Anti-TIM-1 antibody produced in rabbit has been used in immunofluorescence.

Biochem/physiol Actions

Hepatitis A virus cellular receptor 1 (HAVCR1) regulates susceptibility to human immunodeficiency virus-1 (HIV-1) infection. It is upregulated in injured kidneys. HAVCR1 inhibits the activation of the Ga12 G-protein by blocking its association to GTP. Polymorphisms in the protein have been associated with immune dysfunction.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500252.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Tian et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 41(2), 769-783 (2017-02-20)
Kidney injury molecule-1 (KIM-1) is highly expressed in renal tubular cells after injury and is usually regarded as an early biomarker of acute kidney injury(AKI). The aim of this study was to determine the role of KIM-1 in the development
Kidney injury molecule-1 (KIM-1) mediates renal epithelial cell repair via ERK MAPK signaling pathway.
Zhang Z and Cai CX
Molecular and Cellular Biochemistry, 109-116 (2016)
Associations Between TIM1 Polymorphisms and Dilated Cardiomyopathy in a Han Chinese Population.
Xie X
International Heart Journal, 57(6), 742-746 (2016)
Kidney Injury Molecule-1 and Cardiovascular Diseases: From Basic Science to Clinical Practice.
Medic B
BioMed Research International, 854070-854070 (2015)
A 6-amino acid insertion/deletion polymorphism in the mucin domain of TIM-1 confers protections against HIV-1 infection.
Biasin M
Microbes and Infection, 19(1), 69-74 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service