Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2900912

Sigma-Aldrich

Anti-KIAA1324 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

EIG121, Estrogen induced gene 121, KIAA1324, Maba1, RP11-352P4.1, UPF0577 protein KIAA1324

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

western blot: 1-2 μg/mL (Recombinant Protein)
western blot: 5 μg/mL (Cell Lysate)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Synthetic peptide

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebecca Yun et al.
Cell death & disease, 14(8), 546-546 (2023-08-24)
KIAA1324 is a transmembrane protein largely reported as a tumor suppressor and favorable prognosis marker in various cancers, including gastric cancer. In this study, we report the role of N-linked glycosylation in KIAA1324 as a functional post-translational modification (PTM). Loss

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service