Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB2500485

Sigma-Aldrich

Anti-GPX3 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Extracellular glutathione peroxidase, Anti-GPx-P, Anti-GSHPx-3, Anti-GSHPx-P, Anti-Glutathione peroxidase 3 (plasma)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, canine, rat, human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPX3(2878)

General description

Glutathione peroxidase 3 (GPX3) belongs to the glutathione peroxidase (GPX) family. It is located on human chromosome 5q23.

Immunogen

Peptide with sequence C-NQFGKQEPGENSE from the internal region of the protein sequence according to NP_002075.2.

Application

Anti-GPX3 antibody has been used in western blotting.

Biochem/physiol Actions

Glutathione peroxidase 3 (GPX3) blocks oxidative damages by decreasing redundant reactive oxygen species (ROS). It plays a major role in solid tumors and hematological malignancies. GPX3 also plays an important role in the effectiveness of leukemic stem cells (LSCs) and self renewal actions of hematopoietic stem cells (HSCs).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GPX3 methylation in bone marrow predicts adverse prognosis and leukemia transformation in myelodysplastic syndrome
J D Zhou, et al.
Cancer Medicine, 6(1), 267-274 (2017)
Relationship between proinflammatory and antioxidant proteins with the severity of cardiovascular disease in type 2 diabetes mellitus
B G Fontana, et al.
International Journal of Molecular Sciences, 16(5), 9469-9483 (2015)
Beatriz García-Fontana et al.
International journal of molecular sciences, 16(5), 9469-9483 (2015-04-30)
Type 2 diabetes mellitus patients are at significant risk of cardiovascular disease, however, the pathophysiology of these complications is complex and incompletely known in this population. The aim of this study was to compare the serum proteome of patients with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service