Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB2101094

Sigma-Aldrich

Anti-HSP90B1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ECGP, Anti-GP96, Anti-GRP94, Anti-Heat shock protein 90kDa β (Grp94), member 1, Anti-TRA1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

90 kDa

species reactivity

yeast, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSP90B1(7184)

Immunogen

Synthetic peptide directed towards the C terminal region of human HSP90B1

Biochem/physiol Actions

HSP90B1 belongs to the heat shock protein 90 family. It is molecular chaperone that functions in the processing and transport of secreted proteins.

Sequence

Synthetic peptide located within the following region: EEEPEEEPEETAEDTTEDTEQDEDEEMDVGTDEEEETAKESTAEKDEL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abhijna Ballal R et al.
Stem cell research & therapy, 15(1), 108-108 (2024-04-19)
Epilepsy affects ∼60 million people worldwide. Most antiseizure medications in the market act on voltage-gated sodium or calcium channels, indirectly modulating neurotransmitter GABA or glutamate levels or multiple targets. Earlier studies made significant efforts to directly deliver GABA into the brain
Fan Yang et al.
Journal of cellular biochemistry, 119(2), 2427-2439 (2017-09-16)
Mitochondrial dysfunction has emerged as a critical pathophysiological factor of myocardial ischemia/reperfusion (I/R) injury. A thorough understanding of mitochondrial dysfunction during I/R at the molecular level is urgently needed. One prominent microRNA, miR-410, was previously reported to be dynamically regulated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service