Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1412393

Sigma-Aldrich

ANTI-SREBF1 antibody produced in mouse

clone 4G4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

SREBF1, SREBP-1c, SREBP1, bHLHd1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4G4, monoclonal

form

buffered aqueous solution

mol wt

antigen 36.63 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SREBF1(6720)

General description

Sterol regulatory element binding transcription factor 1 (SREBF1) gene is located on human chromosome 17p11.2. SREBF1 encodes the transcription factors SREBP-1a and SREBP-1c. SREBP-1a is expressed in spleen and intestine.SREBP-1c is expressed in liver, adipose tissue and skeletal muscle.

Immunogen

SREBF1 (AAH57388, 801 a.a. ~ 900 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
VTQLFREHLLERALNCVTQPNPSPGSADGDKEFSDALGYLQLLNSCSDAAGAPAYSFSISSSMATTTGVDPVAKWWASLTAVVIHWLRRDEEAAERLCPL

Biochem/physiol Actions

Sterol regulatory element binding transcription factor 1 (SREBF1) regulates lipogenesis, energy homeostasis and insulin sensitivity. It is associated with non-alcoholic fatty liver disease (NAFLD). SREBF1 promotes tumor growth in various cancers. SREBF1 acts as a molecular bridge between lipogenesis and cell cycle control clear cell renal carcinoma.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SREBP-1c as a molecular bridge between lipogenesis and cell cycle progression of clear cell renal carcinoma
Sethi G, et al.
Bioscience Reports, BSR20171270-BSR20171270 (2017)
54G/C polymorphism of SREBF-1 gene is associated with polycystic ovary syndrome
Li L, et al.
European Journal of Obstetrics, Gynecology, and Reproductive Biology, 188, 95-99 (2015)
Lack of association between SREBF-1c gene polymorphisms and risk of non-alcoholic fatty liver disease in a Chinese Han population
Peng XE, et al.
Scientific reports, 6, 32110-32110 (2016)
Association of variants in the sterol regulatory element-binding factor 1 gene (SREBF1) with type 2 diabetes, glycemia, and insulin resistance-A study of 15,734 Danish subjects
Grarup MDN, et al.
Diabetes (2008)
Structure of the human gene encoding sterol regulatory element binding protein-1 (SREBF1) and localization of SREBF1 and SREBF2 to chromosomes 17p11. 2 and 22q13.
Hua X, et al.
Genomics, 25(3), 667-673 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service