Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1410281

Sigma-Aldrich

Anti-CYP27A1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CP27, CTX, CYP27

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 60.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYP27A1(1593)

General description

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This mitochondrial protein oxidizes cholesterol intermediates as part of the bile synthesis pathway. Since the conversion of cholesterol to bile acids is the major route for removing cholesterol from the body, this protein is important for overall cholesterol homeostasis. Mutations in this gene cause cerebrotendinous xanthomatosis, a rare autosomal recessive lipid storage disease. (provided by RefSeq)

Immunogen

CYP27A1 (NP_000775.1, 1 a.a. ~ 531 a.a) full-length human protein.

Sequence
MAALGCARLRWALRGAGRGLCPHGARAKAAIPAALPSDKATGAPGAGPGVRRRQRSLEEIPRLGQLRFFFQLFVQGYALQLHQLQVLYKAKYGPMWMSYLGPQMHVNLASAPLLEQVMRQEGKYPVRNDMELWKEHRDQHDLTYGPFTTEGHHWYQLRQALNQRLLKPAEAALYTDAFNEVIDDFMTRLDQLRAESASGNQVSDMAQLFYYFALEAICYILFEKRIGCLQRSIPEDTVTFVRSIGLMFQNSLYATFLPKWTRPVLPFWKRYLDGWNAIFSFGKKLIDEKLEDMEAQLQAAGPDGIQVSGYLHFLLASGQLSPREAMGSLPELLMAGVDTTSNTLTWALYHLSKDPEIQEALHEEVVGVVPAGQVPQHKDFAHMPLLKAVLKETLRLYPVVPTNSRIIEKEIEVDGFLFPKNTQFVFCHYVVSRDPTAFSEPESFQPHRWLRNSQPATPRIQHPFGSVPFGYGVRACLGRRIAELEMQLLLARLIQKYKVVLAPETGELKSVARIVLVPNKKVGLQFLQRQC

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service