Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-
biological source
rabbit
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 83.1 kDa
species reactivity
human
technique(s)
western blot: 1 μg/mL
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... STON1(11037)
General description
Endocytosis of cell surface proteins is mediated by a complex molecular machinery that assembles on the inner surface of the plasma membrane. This gene encodes one of two human homologs of the Drosophila melanogaster stoned B protein. This protein is related to components of the endocytic machinery and exhibits a modular structure consisting of an N-terminal proline-rich domain, a central region of homology specific to the human stoned B-like proteins, and a C-terminal region homologous to the mu subunits of adaptor protein (AP) complexes. Co-transcription of this gene and the neighboring downstream gene generates a rare transcript (SALF), which encodes a fusion protein comprised of sequence sharing identity with each individual gene product. (provided by RefSeq)
Immunogen
STON1 (NP_006864.2, 1 a.a. ~ 735 a.a) full-length human protein.
Sequence
MCSTNPGKWVTFDDDPAVQSSQKSKNFPLENQGVCRPNGLKLNLPGLREFPSGSSSTSSTPLSSPIVDFYFSPGPPSNSPLSTPTKDFPGFPGIPKAGTHVLYPIPESSSDSPLAISGGESSLLPTRPTCLSHALLPSDHSCTHPTPKVGLPDEVNPQQAESLGFQSDDLPQFQYFREDCAFSSPFWKDEGSDSHFTLDPPGSKKMFSSRNKEMPIDQKSLNKCSLNYICEKLEHLQSAENQDSLRSLSMHCLCAEENASSFVPHTLFRSQPKSGWSFMLRIPEKKNMMSSRQWGPIFLKVLPGGILQMYYEQGLEKPFKEIQLDPYCRLSEPKVENFSVAGKIHTVKIEHVSYTEKRKYHSKTEVVHEPDIEQMLKLGSTSYHDFLDFLTTVEEELMKLPAVSKPKKNYEEQEISLEIVDNFWGKVTKEGKFVESAVITQIYCLCFVNGNLECFLTLNDLELPKRDESYYEKDSEKKGIDILDYHFHKCVNVQEFEQSRIIKFVPLDACRFELMRFKTLYNGDNLPFSLKSVVVVQGAYVELQAFVNMASLAQRSSYAGSLRSCDNIRIHFPVPSQWIKALWTMNLQRQKSLKAKMNRRACLGSLQELESEPVIQVTVGSAKYESAYQAVVWKIDRLPDKNSSLDHPHCLSYKLELGSDQEIPSDWYPFATVQFSVPDTCASRTEVRSLGVESDVQPQKHVQQRACYNIQVEIEKKWIKIDGEDPDKIGDCITQ
Sequence
MCSTNPGKWVTFDDDPAVQSSQKSKNFPLENQGVCRPNGLKLNLPGLREFPSGSSSTSSTPLSSPIVDFYFSPGPPSNSPLSTPTKDFPGFPGIPKAGTHVLYPIPESSSDSPLAISGGESSLLPTRPTCLSHALLPSDHSCTHPTPKVGLPDEVNPQQAESLGFQSDDLPQFQYFREDCAFSSPFWKDEGSDSHFTLDPPGSKKMFSSRNKEMPIDQKSLNKCSLNYICEKLEHLQSAENQDSLRSLSMHCLCAEENASSFVPHTLFRSQPKSGWSFMLRIPEKKNMMSSRQWGPIFLKVLPGGILQMYYEQGLEKPFKEIQLDPYCRLSEPKVENFSVAGKIHTVKIEHVSYTEKRKYHSKTEVVHEPDIEQMLKLGSTSYHDFLDFLTTVEEELMKLPAVSKPKKNYEEQEISLEIVDNFWGKVTKEGKFVESAVITQIYCLCFVNGNLECFLTLNDLELPKRDESYYEKDSEKKGIDILDYHFHKCVNVQEFEQSRIIKFVPLDACRFELMRFKTLYNGDNLPFSLKSVVVVQGAYVELQAFVNMASLAQRSSYAGSLRSCDNIRIHFPVPSQWIKALWTMNLQRQKSLKAKMNRRACLGSLQELESEPVIQVTVGSAKYESAYQAVVWKIDRLPDKNSSLDHPHCLSYKLELGSDQEIPSDWYPFATVQFSVPDTCASRTEVRSLGVESDVQPQKHVQQRACYNIQVEIEKKWIKIDGEDPDKIGDCITQ
Physical form
Solution in phosphate buffered saline, pH 7.4
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.