Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB1406579

Sigma-Aldrich

Anti-VCAM1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CD106, DKFZp779G2333, INCAM-100, MGC99561

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~81.3 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VCAM1(7412)

General description

This gene is a member of the Ig superfamily and encodes a cell surface sialoglycoprotein expressed by cytokine-activated endothelium. This type I membrane protein mediates leukocyte-endothelial cell adhesion and signal transduction, and may play a role in the development of artherosclerosis and rheumatoid arthritis. Two alternatively spliced transcripts encoding different isoforms have been described for this gene. (provided by RefSeq)

Vascular cell adhesion molecule 1 (VCAM1) is activated by transforming growth factor β-1 (TGF-β1) and is part of the immunoglobulin superfamily. The protein is expressed in blood vessels. VCAM1 possesses many extracellular immunoglobulin-like domains, a transmembrane region and a cytoplasmic domain. The VCAM1 gene is localized to human chromosome 1p21.2.

Immunogen

VCAM1 (NP_001069.1, 1 a.a. ~ 739 a.a) full-length human protein.

Sequence
MPGKMVVILGASNILWIMFAASQAFKIETTPESRYLAQIGDSVSLTCSTTGCESPFFSWRTQIDSPLNGKVTNEGTTSTLTMNPVSFGNEHSYLCTATCESRKLEKGIQVEIYSFPKDPEIHLSGPLEAGKPITVKCSVADVYPFDRLEIDLLKGDHLMKSQEFLEDADRKSLETKSLEVTFTPVIEDIGKVLVCRAKLHIDEMDSVPTVRQAVKELQVYISPKNTVISVNPSTKLQEGGSVTMTCSSEGLPAPEIFWSKKLDNGNLQHLSGNATLTLIAMRMEDSGIYVCEGVNLIGKNRKEVELIVQEKPFTVEISPGPRIAAQIGDSVMLTCSVMGCESPSFSWRTQIDSPLSGKVRSEGTNSTLTLSPVSFENEHSYLCTVTCGHKKLEKGIQVELYSFPRDPEIEMSGGLVNGSSVTVSCKVPSVYPLDRLEIELLKGETILENIEFLEDTDMKSLENKSLEMTFIPTIEDTGKALVCQAKLHIDDMEFEPKQRQSTQTLYVNVAPRDTTVLVSPSSILEEGSSVNMTCLSQGFPAPKILWSRQLPNGELQPLSENATLTLISTKMEDSGVYLCEGINQAGRSRKEVELIIQVTPKDIKLTAFPSESVKEGDTVIISCTCGNVPETWIILKKKAETGDTVLKSIDGAYTIRKAQLKDAGVYECESKNKVGSQLRSLTLDVQGRENNKDYFSPELLVLYFASSLIIPAIGMIIYFARKANMKGSYSLVEAQKSKV

Biochem/physiol Actions

Vascular cell adhesion molecule 1 (VCAM1) plays a role in the adhesion of monocytes, lymphocytes, basophils and eosinophils to the vascular endothelium. It also functions in signal transduction and adhesion of leukocytes to endothelial cells. The protein has been associated with atherosclerosis and rheumatoid arthritis. VCAM1 is upregulated in idiopathic pulmonary fibrosis. It has been linked to cancer progression and metastasis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

VCAM-1 is a TGF-β1 inducible gene upregulated in idiopathic pulmonary fibrosis.
Agassandian M, et al.
Cellular Signalling, 27(12), 2467-2473 (2015)
Vascular cell adhesion molecule-1 (VCAM-1)?An increasing insight into its role in tumorigenicity and metastasis.
Schlesinger M and Gerd B
International Journal of Cancer. Journal International Du Cancer, 136(11), 2504-2514 (2015)
Pharmacogenomics of multigenic diseases: sex-specific differences in disease and treatment outcome.
Pinsonneault J and Wolfgang S
AAPS PharmSci, 5(4), 49-61 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service