Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

SAB1404621

Sigma-Aldrich

Monoclonal Anti-RIPK2, (C-terminal) antibody produced in mouse

clone 6F7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

CARD3, CARDIAK, CCK, GIG30, RICK, RIP2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6F7, monoclonal

form

buffered aqueous solution

mol wt

antigen ~38.21 kDa

species reactivity

mouse, human, rat

technique(s)

capture ELISA: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RIPK2(8767)

Related Categories

General description

Receptor interacting serine/threonine kinase 2 (RIPK2) belongs to the RIP kinase family. The protein contains a caspase activation and recruitment domain (CARD) at the C-terminal, N-terminal kinase domain, and a bridging intermediate domain. The gene is mapped to human chromosome 8q21.3.
This gene encodes a member of the receptor-interacting protein (RIP) family of serine/threonine protein kinases. The encoded protein contains a C-terminal caspase activation and recruitment domain (CARD), and is a component of signaling complexes in both the innate and adaptive immune pathways. It is a potent activator of NF-kappaB and inducer of apoptosis in response to various stimuli. (provided by RefSeq)

Immunogen

RIPK2 (AAH04553, 431 a.a. ~ 540 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
LQPGIAQQWIQSKREDIVNQMTEACLNQSLDALLSRDLIMKEDYELVSTKPTRTSKVRQLLDTTDIQGEEFAKVIVQKLKDNKQMGLQPYPEILVVSRSPSLNLLQNKSM

Application

Monoclonal Anti-RIPK2, (C-terminal) antibody produced in mouse has been used in immunoblotting (1:500).

Biochem/physiol Actions

Receptor interacting serine/threonine kinase 2 (RIPK2) is a key regulator of the immune and inflammatory pathways. It is a potent activator of nuclear factor (NF)-κB via nucleotide-binding and oligomerization domain (NOD) receptor. RIPK2 is associated with the progression and aggressiveness, tumor size, metastasis, and overall stagging in various types of cancers. Overexpression of RIPK2 is observed in the head and neck squamous cell carcinoma (HNSCC), gastric cancer, colorectal cancer (CRC), and lethal prostate cancers. It is found to induce cell proliferation and inhibit apoptosis in glioma and breast cancer. RIPK2 polymorphism is also associated with the onset of bladder cancer.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vivek Misra
Annals of neurosciences, 21(2), 69-73 (2014-09-11)
Available research data in Autism suggests the role of a network of brain areas, often known as the 'social brain'. Recent studies highlight the role of genetic mutations as underlying patho-mechanism in Autism. This mini review, discusses the basic concepts
Rola F Jaafar et al.
Medicina (Kaunas, Lithuania), 57(7) (2021-08-07)
Background and objectives: Receptor-interacting serine/threonine-protein kinase-2 (RIPK2) is an important mediator in different pathways in the immune and inflammatory response system. RIPK2 was also shown to play different roles in different cancer types; however, in colorectal cancer (CRC), its role
Ueli Nachbur et al.
Nature communications, 6, 6442-6442 (2015-03-18)
Intracellular nucleotide binding and oligomerization domain (NOD) receptors recognize antigens including bacterial peptidoglycans and initiate immune responses by triggering the production of pro-inflammatory cytokines through activating NF-κB and MAP kinases. Receptor interacting protein kinase 2 (RIPK2) is critical for NOD-mediated
Lucien P Garo et al.
Nature communications, 12(1), 2419-2419 (2021-04-25)
Chronic inflammation can drive tumor development. Here, we have identified microRNA-146a (miR-146a) as a major negative regulator of colonic inflammation and associated tumorigenesis by modulating IL-17 responses. MiR-146a-deficient mice are susceptible to both colitis-associated and sporadic colorectal cancer (CRC), presenting
Yongyu Chen et al.
Theranostics, 10(1), 323-339 (2020-01-07)
Aims: We aimed to measure the abundance of Fusobacterium nucleatum (F. nucleatum) in colorectal cancer (CRC) tissues from patients and to uncover the function of this bacterium in colorectal tumor metastasis. Methods: We collected metastatic and non-metastatic CRC tissues to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service