Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1401190

Sigma-Aldrich

Anti-IAPP antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

AMYLIN, DAP, IAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IAPP(3375)

General description

Islet, or insulinoma, amyloid polypeptide is commonly found in pancreatic islets of patients suffering diabetes mellitus type II, or harboring an insulinoma. While the assosciation of amylin with the development of type II diabetes has been known for some time, a direct causative role for amylin has been harder to establish. Studies suggest that amylin, like the related beta-amyloid (Abeta) associated with Alzheimer′s disease, can induce apoptotic cell-death in particular cultured cells, an effect that may be relevant to the development of type II diabetes. (provided by RefSeq)

Immunogen

IAPP (NP_000406.1, 1 a.a. ~ 89 a.a) full-length human protein.

Sequence
MGILKLQVFLIVLSVALNHLKATPIESHQVEKRKCNTATCATQRLANFLVHSSNNFGAILSSTNVGSNTYGKRNAVEVLKREPLNYLPL

Biochem/physiol Actions

IAPP (islet amyloid polypeptide) is secreted by the pancreatic β-cells along with insulin. IAPP is known to be involved in the regulation of gastric emptying, satiety and inhibiting glucagon secretion. IAPP is associated with type 2 diabetes mellitus where IAPP is part of amyloid deposits and is associated with mass and functional loss of β-cells. Oligomers and fibrils formed by IAPP is known to be toxic to the pancreatic islet β-cells.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of Aluminium and EGCG on Fibrillation and Aggregation of Human Islet Amyloid Polypeptide.
Xu ZX
Journal of Diabetes Research, 2016:1867059, 1-14 (2016)
Disease-linked mutations in factor H reveal pivotal role of cofactor activity in self-surface-selective regulation of complement activation.
Kerr H
The Journal of Biological Chemistry, 292(32), 13345-13360 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service