Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

SAB1400262

Sigma-Aldrich

Anti-SLC3A2 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-4F2, Anti-4F2HC, Anti-4T2HC, Anti-CD98, Anti-CD98HC, Anti-MDU1, Anti-NACAE

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC3A2(6520)

General description

This gene is a member of the solute carrier family and encodes a cell surface, transmembrane protein with an alpha amylase domain. The protein exists as the heavy chain of a heterodimer, covalently bound through di-sulfide bonds to one of several possible light chains. It associates with integrins and mediates integrin-dependent signaling related to normal cell growth and tumorigenesis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. (provided by RefSeq)
Solute carrier family 3 member 2 (SLC3A2) is a type II transmembrane glycoprotein which is widely expressed. It is expressed on the cell membrane and in some types of endosomes. The protein is part of the heterodimeric amino acid transporter family. The SLC3A2 gene is localized on human chromosome 11q13.

Immunogen

SLC3A2 (NP_001013269.1, 1 a.a. ~ 529 a.a) full-length human protein.

Sequence
MSQDTEVDMKEVELNELEPEKQPMNAASGAAMSLAGAEKNGLVKIKVAEDEAEAAAAAKFTGLSKEELLKVAGSPGWVRTRWALLLLFWLGWLGMLAGAVVIIVRAPRCRELPAQKWWHTGALYRIGDLQAFQGHGAGNLAGLKGRLDYLSSLKVKGLVLGPIHKNQKDDVAQTDLLQIDPNFGSKEDFDSLLQSAKKKSIRVILDLTPNYRGENSWFSTQVDTVATKVKDALEFWLQAGVDGFQVRDIENLKDASSFLAEWQNITKGFSEDRLLIAGTNSSDLQQILSLLESNKDLLLTSSYLSDSGSTGEHTKSLVTQYLNATGNRWCSWSLSQARLLTSFLPAQLLRLYQLMLFTLPGTPVFSYGDEIGLDAAALPGQPMEAPVMLWDESSFPDIPGAVSANMTVKGQSEDPGSLLSLFRRLSDQRSKERSLLHGDFHAFSAGPGLFSYIRHWDQNERFLVVLNFGDVGLSAGLQASDLPASASLPAKADLLLSTQPGREEGSPLELERLKLEPHEGLLLRFPYAA

Biochem/physiol Actions

Solute carrier family 3 member 2 (SLC3A2) has a role in amino acid transport. It associates with integrins and modulates their signaling functions. The protein is involved in cell-to-cell fusion occurring between macrophages or trophoblasts. SLC3A2 is overexpressed in various cancers.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CD98hc (SLC3A2) drives integrin-dependent renal cancer cell behavior.
Poettler M
Molecular Cancer (2013)
Brucella Intracellular Life Relies on the Transmembrane Protein CD98 Heavy Chain.
Keriel A
The Journal of Infectious Diseases (2015)
Development of Human Membrane Transporters: Drug Disposition and Pharmacogenetics
Miriam G
Clinical Pharmacy (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service